ContactUsLinkComponent
Water baths, WTB series
Water baths, WTB series
Supplier:  MEMMERT
Certificates
undefined
Water baths, WTB series
Supplier:  MEMMERT
Specifications
  • Catalog No:
  • 462-0614
  • 462-0615
  • 462-0611
  • 462-0612
  • 462-0613
  • 462-0610
  • Capacity:
  • 51 L
  • 7,5 L
  • 17, 1 L
  • 23,1 L
  • 37,5 L
  • 10 L
  • Temperature range:
  • RT* +5...100 °C
  • RT* +5...100 °C
  • RT* +5...100 °C
  • RT* +5...100 °C
  • RT* +5...100 °C
  • RT* +5...100 °C
  • Timer:
  • Digital timer from 1 min. up to 99 hours 59 min
  • Digital timer from 1 min. up to 99 hours 59 min
  • Digital timer from 1 min. up to 99 hours 59 min
  • Digital timer from 1 min. up to 99 hours 59 min
  • Digital timer from 1 min. up to 99 hours 59 min
  • Digital timer from 1 min. up to 99 hours 59 min
  • Heating capacity:
  • 2800 W
  • 1000 W
  • 1420 W
  • 1420 W
  • 2800 W
  • 1000 W
  • Shipping weight:
  • 23 kg
  • 10 kg
  • 14 kg
  • 16 kg
  • 20 kg
  • 12 kg
Specifications
Related Information
Peltier-cooling device for waterbaths
Product Information
Frequently Bought Together