
Penicillin/Streptomycin/Neomycin solution (~5,000 units penicillin, 5 mg streptomycin and 10 mg neomycin/ml) stabilizovaný, Bioreagent cell culture reagent, sterile filtered
Supplier: Merck
Certificates
About this item
:
:M-Clarity™ quality level = MQ200
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance (Form):Liquid
- Appearance (Turbidity) :Clear
- Sterility:Pass
- Endotoxin Level at 10 ml/L:< 0.5 EU/ml
- Cell Line:Hep-2 Human Cells
- Cell Culture Test (percent of control):> 50 %
Specification Test Results
Frequently Bought Together