Order Entry
Czech Republic
ContactUsLinkComponent
Sodium dihydrogen phosphate, anhydrous ≥99%, HiPerSolv CHROMANORM® for HPLC
Sodium dihydrogen phosphate, anhydrous ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates
undefined
Sodium dihydrogen phosphate, anhydrous ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    ≥ 99 %
  • Appearance:
    White crystalline powder
  • Filter test:
    Conforms to standard
  • RP gradient test :
    Conforms to standard
  • Solubility in water (0.1 g/ml; 25°C):
    Colourless, complete
  • pH (20°C; 10 %):
    4,0 - 4,5
  • Absorbance (210 nm) (0.5 mol/l):
    ≤ 0,1
  • Absorbance (260 nm) (0.5 mol/l):
    ≤ 0,06
  • Absorbance (280 nm) (0.5 mol/l):
    ≤ 0,04
  • Absorbance (320 nm) (0.5 mol/l):
    ≤ 0,02
  • Absorbance (500 nm) (0.5 mol/l):
    ≤ 0,02

Specification Test Results

Related Information

Frequently Bought Together