ContactUsLinkComponent
Nitrite test strips, Dosatest
Nitrite test strips, Dosatest
  85705.601
 :  
undefined
Nitrite test strips, Dosatest
  85705.601
 :  
  • Chemical method:
    Griess' reaction
  • Pk:
    100
  • Description:
    Nitrite test strips
  • Tests per kit:
    100
  • Range:
    0 • 0.1 • 0.3 • 0.6 • 1 • 2 • 3 g/L NO₂⁻
Semi-quantitative test strips for the determination of nitrite.
  • Ideal for rapid and easy analysis of nitrite directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to red
 : Delivered as 100 test strips in a tube, instruction leaflet in a box
 : Strongly acidic solutions must be diluted with dilute sodium hydroxide and strongly alkaline samples with dilute sulphuric acid to pH 4 to 6.