Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluoro-Jade
Fluoro-Jade B (FJB), powder for identifying Degenerating Neurons
Fluoro-Jade B (FJB), powder for identifying Degenerating Neurons
Fluoro-Jade B (FJB), powder for identifying Degenerating Neurons
Catalog # BSENTR-150-FJB
Supplier:
Biosensis
CAS Number:
Expand image
Fluoro-Jade B (FJB), powder for identifying Degenerating Neurons
Catalog # BSENTR-150-FJB
Supplier:
Biosensis
CAS Number:
Specifications
Pack type:
Glass bottle
Pk:
30 mg
Seq:
2345
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
Product Details
Documentation