
p-Toluenesulphonic acid monohydrate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:6192-52-5
:
Formula:C₇H₈O₃S·1H₂O
MDL Number:MFCD00142137
:190.22 g/mol
Merck Index:12,09671
Melting Pt:102…104 °C
Density:1.24 g/cm³ (20 °C)
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:203-180-0
UN:2585
ADR:8,III
:GHS07
Specification Test Results
- Assay:≥ 98 %
- White deliquescent crystalline powder:Passes test
- Water:About 11 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87098.180 spec_VWRC87098.180 spec_VWRC87098.290 spec_VWRC87098.460
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together