Globe stirrer couplings, BOLA
Dodavatel: Bohlender
Certifikáty
O tomto produktu
These POM couplings are ideal for balancing misalignment of axes between stirrer and stirrer shaft. Suitable for glass, metal or Bola stirrer shafts. Both models are suitable for rotational speeds of up to 1200 rpm and can be used for both right and left-handed rotation.
- Excellent chemical resistance, temperature resistance from –20 to +110 °C
- Assembled by means of screw joints with clamp rings
- Pivot (length 90 mm) can be shortened by the user
- Maximum misalignment of 10 mm
Powerful transmission of up to 300 Ncm.
Powerful transmission of up to 300 Ncm.
Frequently Bought Together