Flame-retardant bomber jackets, high visibility, Bizflame™ Rain, S773
: Portwest
These fully waterproof anti-static bomber jackets provide multi-norm protection in hazardous conditions. These are designed with chemical resistance and a detachable hood.
- Flame resistant treated waterproof fabric prevents water penetration
- Premium sew on flame resistant reflective tape
- Fully lined and padded to trap the heat and increase warmth
- Taped seams to provide additional protection
- FR cotton lined for added warmth and comfort
- 4 pockets for ample storage
- Phone pocket
Designed with breathable fabric to draw moisture away from the body keeping the wearer cool, dry and comfortable.
Bizflame™ Rain material is combining a 98% polyester and 2% anti-static carbon fibre (250 g), this breathable fabric offers maximum protection to a range of International standards.
Shell fabric: Bizflame® Rain (250 g)
Lining fabric: 100% cotton flame resistant lining (170 g)
Filling fabric: 100% polyester flame resistant filling (100 g)
: ANSI/ISEA 107: 2015 TYPE R CLASS 3; EN ISO 14116 Index 3; EN 1149 -5; RIS-3279-TOM ISSUE 1 (ORANGE ONLY); EN 13034 Type 6;
EN ISO 20471 Class 3; EN 343 Class 3:1
EN ISO 20471 Class 3; EN 343 Class 3:1
: Use non-biological powder. Do not use softener.
- Catalog No:
- PRTWS773ORRL
- PRTWS773ORRM
- PRTWS773ORRXXL
- PRTWS773YERM
- PRTWS773YERS
- PRTWS773ORRXL
- PRTWS773YERXL
- PRTWS773YERL
- PRTWS773ORRXXXL
- PRTWS773YERXXXL
- PRTWS773YERXXL
- PRTWS773YER5XL
- PRTWS773YER4XL
- PRTWS773ORRS
- Model:
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- S773
- Garment application:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- 98% polyester and 2% anti-static carbon fibre (250 g)
- Weight:
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
- 250 g
Frequently Bought Together