ContactUsLinkComponent
Flexible fit probed test earplugs, E-A-R™
Flexible fit probed test earplugs, E-A-R™
Catalog # MMMA393-2026-50
Supplier:  3M
undefined
Flexible fit probed test earplugs, E-A-R™
Catalog # MMMA393-2026-50
Supplier:  3M
Specifications
  • Description:
    Flexible fit probed test earplugs
  • Pk:
    50 PAIR
Specifications
About this item
E-A-R™ Flexible Fit Earplugs equipped with a probe tube for fit testing using the 3M™ E-A-Rfit™ Dual-Ear Validation System. Cannot be used for hearing protection.
  • Test earplugs are modified with a probe tube for use during earplug fit testing
  • 50 pairs per case
  • For occupational use only
These test Plugs are a specially probed version of the standard earplug. This product works with the 3M™ E-A-Rfit™ Dual-Ear Validation System to allow fit tests of Flexible Fit earplugs.