Analytical standard, Myclobutanil, reference material, Fluka™
Catalog # HONCR1760-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:88671-89-0
- Description:Myclobutanil
- Description:Myclobutanil
- Form:Neat