Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, lambda-Cyhalothrin, reference material, Fluka™
  HONCR1756-100MG
 :  
Analytical standard, lambda-Cyhalothrin, reference material, Fluka™
  HONCR1756-100MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    91465-08-6
  • Description:
    lambda-Cyhalothrin
  • Description:
    lambda-Cyhalothrin
  • Form:
    Neat