Analytical standard, Aminoethoxyvinyl glycine hydrochloride, reference material, Fluka™
Catalog # HONCR1747-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- Standards:Reference material
- CAS No.:55720-26-8
- Description:Aminoethoxyvinyl glycine hydrochloride
- Storage temperature:–20 °C
- Description:Aminoethoxyvinyl glycine hydrochloride
- Form:Neat