Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Bupirimate, reference material, Fluka™
Catalog # HONCR1718-100MG
CAS Number:  
Analytical standard, Bupirimate, reference material, Fluka™
Catalog # HONCR1718-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    41483-43-6
  • Description:
    Bupirimate
  • Storage temperature:
    2…8 °C
  • Description:
    Bupirimate
  • Form:
    Neat

Specifications

About this item