Order Entry
Czech Republic
ContactUsLinkComponent
Analytical standard, Malathion, reference material, Fluka™
Analytical standard, Malathion, reference material, Fluka™
Catalog # HONCR1782-100MG
Dodavatel:  Honeywell Chemicals
Číslo CAS:  
Analytical standard, Malathion, reference material, Fluka™
Catalog # HONCR1782-100MG
Dodavatel:  Honeywell Chemicals
Číslo CAS:  

Specifikace

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    121-75-5
  • Description:
    Malathion
  • Storage temperature:
    2…8 °C
  • Description:
    Malathion
  • Form:
    Neat

Specifikace

O tomto produktu