Orders LinkContactUsLinkComponent
Beryllium AAS Standards, (CRMs), Fluka™
Supplier:   Honeywell Chemicals
Beryllium AAS Standards, (CRMs), Fluka™
Certificates
Beryllium AAS Standards, (CRMs), Fluka™
3 Options Available Below
Product Details & Documents
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
 : ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Product Family Options
Product Information
  • Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Beryllium
    Be
    1000 mg/L
    2% HCl
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 4 200,00 Kč
    4 200,00 Kč
    1 * 100 mL
       4 200,00 Kč
  • Beryllium
    Be
    1000 mg/L
    2% HNO₃/1% HF
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 4 300,00 Kč
    4 300,00 Kč
    1 * 100 mL
       4 300,00 Kč
  • Beryllium
    Be
    1000 mg/L
    2% HNO₃/1% HF
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 9 380,00 Kč
    9 380,00 Kč
    1 * 500 mL
       9 380,00 Kč