3-Methoxypropylamine Standard for Ion Chromatography, (CRM), Fluka™
HONCE3028-100ML
:
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:3-Methoxypropylamine standard
Certified Reference Material (CRM)
- Traceable to SRM from NIST
: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).