Beryllium ICP & ICP-MS Standard, (CRMs), Fluka™
Catalog # HONCE1375-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:2% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Beryllium
- Symbol:Be
Specifications
About this item
Certified Reference Material (CRM)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).