Order Entry
Czech Republic
ContactUsLinkComponent
Cadmium ICP standards, (CRMs), Fluka™
Cadmium ICP standards, (CRMs), Fluka™
Catalog # HONCE1139-100ML
CAS Number:  
Cadmium ICP standards, (CRMs), Fluka™
Catalog # HONCE1139-100ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).