Order Entry
Czech Republic
ContactUsLinkComponent
Internal standard mix for ICP-MS systems
Internal standard mix for ICP-MS systems
Catalog # HONCE2201-100ML
CAS Number:  
Internal standard mix for ICP-MS systems
Catalog # HONCE2201-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Internal Standard Mix

Specifications

About this item

Certified Reference Material (CRM).

50 mg/L Sc, 20 mg/L Ge, 10 mg/L each of In, Ir, 6Li, Rh, Tb, Y in HNO₃ 2%