Order Entry
Czech Republic
ContactUsLinkComponent
 
Combined Six Cation Standard I for IC
Catalog # HONCE2032-100ML
CAS Number:  
Combined Six Cation Standard I for IC
Catalog # HONCE2032-100ML
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Combined Six Cation Standard I

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L Ca2+, 400 mg/L NH4+, 200 mg/L each of Na+, K+, Mg2+, 50 mg/L Li+ Material in H₂O