Combined Five Anion Standard for IC
Catalog # HONCE2031-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Combined Five Anion Standard
Specifications
About this item
Certified Reference Material (CRM).
150 mg/L each of PO43-, SO42-, 100 mg/L NO3-, 30 mg/L Cl-, 20 mg/L F- in H₂O