Nickel (II) nitrate 10 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™
Catalog # HONCE4605-50ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Matrix:HNO₃ 1%
- Concentration:Ni(NO₃)₂ 10,000 mg/L
- Quality:ISO/IEC17025:2017(ANAB),ISO17034:2016(ANAB)
- Form:Solution
- Density:1.01g/cm3
Specification Test Results
About this item
:
:182.7 g/mol
Density:1.01 g/cm3
Storage Temperature:Ambient
CAS Number:13138-45-9
Formula:N₂NiO₆
MDL Number:MFCD00011139