Order Entry
Czech Republic
ContactUsLinkComponent
Corning® mouse laminin/entactin complex, high concentration (from Engelbreth-Holm-Swarm mouse tumour)
Corning® mouse laminin/entactin complex, high concentration (from Engelbreth-Holm-Swarm mouse tumour)
Catalog # 734-1273
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
undefined
Corning® mouse laminin/entactin complex, high concentration (from Engelbreth-Holm-Swarm mouse tumour)
Catalog # 734-1273
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    10,5 mg
  • Source:
    Engelbreth-Holm-Swarm mouse tumour
  • Species:
    Mouse
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Storage conditions:
    Stable for at least three months at −70 °C. Do not store in frost-free freezer. Avoid multiple freeze-thaws.
  • Environmental certification:
    ISO 14001 - Bedford
  • Protein/peptide name:
    Laminin/entactin complex, high concentration
  • Formulation:
    ≥10 mg/ml Phosphate-Buffered Saline

Specifications

About this item

Laminin, a major component of basement membranes, has numerous biological activities including promotion of cell adhesion, migration, growth, and differentiation, including neurite outgrowth. It can be used as a thin coating on tissue culture surfaces or as a soluble additive to culture medium. The Laminin/Entactin Complex, high concentration (HC) is a special formulation that has been developed for three-dimensional (3D) culture.

Quality controlled by SDS-PAGE and tested and found negative for bacteria, fungi, and mycoplasma.