ContactUsLinkComponent
X-ray and metal detectable long handle scoops
X-ray and metal detectable long handle scoops
Catalog # SAMPM1040B-100
Dodavatel:  Sampling Systems
undefined
X-ray and metal detectable long handle scoops
Catalog # SAMPM1040B-100
Dodavatel:  Sampling Systems
Specifikace
  • Sterile:
    -
  • Capacity:
    100 ml
  • Colour:
    Blue
  • Pk:
    4
Specifikace
O tomto produktu
These long handled metal detectable scoops are perfect for both sampling and dispensing when working with powders and granules.
  • Robust PP construction gives excellent chemical resistance to many products
  • Supplied Individually bagged and produced in a cleanroom
  • Full batch traceability
The long handle and the bright blue colour make the scoops easy to spot if they get left behind. They are both X-ray and metal detectable.
To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom. The robust nature of these scoops means that they can give many years of service or for more demanding applications they can be used as a single use disposable product straight from the bag.
Certifications: These scoops conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.