ContactUsLinkComponent
Avantor® Hichrom HI-SOLVE 2, GC Columns
Avantor® Hichrom HI-SOLVE 2, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-SOLVE 2, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI63-25-025-30
  • HI63-32-025-50
  • Phase name:
  • HI-SOLVE 2
  • HI-SOLVE 2
  • Phase composition:
  • Proprietary Phase developed for complex solvent mix
  • Proprietary Phase developed for complex solvent mix
  • Crossbond:
  • Stabilised
  • Stabilised