ContactUsLinkComponent
Avantor® Hichrom HI-35 HT, GC Columns
Avantor® Hichrom HI-35 HT, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-35 HT, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI89-32-015-30
  • HI89-25-010-15
  • HI89-32-025-15
  • HI89-53-025-30
  • HI89-25-015-30
  • HI89-53-010-15
  • HI89-25-025-15
  • HI89-53-010-30
  • HI89-25-010-30
  • HI89-32-025-30
  • HI89-25-015-15
  • HI89-53-010-10
  • HI89-53-017-30
  • HI89-32-010-15
  • HI89-10-010-5
  • HI89-53-025-15
  • HI89-32-010-30
  • HI89-53-015-15
  • HI89-25-025-30
  • HI89-32-010-10
  • HI89-10-010-10
  • HI89-53-015-30
  • HI89-32-015-15
  • Phase name:
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • HI-35 HT
  • Phase composition:
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • 35% Phenyl, 65% Methyl Polysiloxane - high temperature
  • Polarity:
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Mid-polarity stationary phase
  • Crossbond:
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
  • Temperature limit:
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • USP code:
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42
  • G28, G32, G42