ContactUsLinkComponent
Avantor® Hichrom HI-DEX G-03, GC Columns
 :  Avantor
undefined
Avantor® Hichrom HI-DEX G-03, GC Columns
 :  Avantor
  • Catalog No:
  • HI75-25-015-15
  • HI75-25-015-25
  • HI75-25-015-60
  • HI75-25-025-15
  • HI75-25-015-30
  • HI75-25-015-50
  • HI75-25-025-50
  • HI75-25-025-60
  • HI85-10-010-10
  • HI75-25-025-25
  • HI75-25-025-30
  • HI85-10-010-5
  • Phase name:
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • HI-DEX G-03
  • Phase composition:
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Proprietary Cyclodexstrin based
  • Polarity:
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Crossbond:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Temperature limit:
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
  • 250 °C
Frequently Bought Together