ContactUsLinkComponent
Avantor® Hichrom HI-PAH 2, GC Columns
Avantor® Hichrom HI-PAH 2, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-PAH 2, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HIGCHI49-25-025-30
  • HIGCHI49-15-010-15
  • HI49-18-010-40
  • HI49-18-010-20
  • HI49-25-010-60
  • HI49-25-010-30
  • Phase name:
  • HI-PAH 2
  • HI-PAH 2
  • HI-PAH 2
  • HI-PAH 2
  • HI-PAH 2
  • HI-PAH 2
  • Phase composition:
  • Proprietary Phase developed for EU PAHs analysis
  • Proprietary Phase developed for EU PAHs analysis
  • Proprietary Phase developed for EU PAHs analysis
  • Proprietary Phase developed for EU PAHs analysis
  • Proprietary Phase developed for EU PAHs analysis
  • Proprietary Phase developed for EU PAHs analysis
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C
  • 350 °C