Avantor® Hichrom HI-1 MS, GC Columns
Supplier: Avantor
Certificates
About this item
Really versatile general purpose apolar phase for a wide range of MS applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semivolatiles, oxygenates. HI-1 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
- Phase composition: Crossbonded 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - MS grade
- Long lifetime
- Equivalent to USP phases G1, G2, G38
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI16-32-025-50
- HI16-32-025-30
- HI16-18-018-40
- HI16-20-040-50
- HI16-32-050-30
- HI16-18-018-10
- HI16-32-050-105
- HI16-20-040-20
- HI16-20-100-20
- HI16-53-150-50
- HI16-32-100-10
- HI16-20-100-40
- HI16-32-050-60
- HI16-25-010-15
- HI16-10-010-10
- HI16-05-005-2.5
- HI16-53-150-10
- HI16-53-100-60
- HI16-25-100-60
- HIGCHI1002502515
- HI16-20-020-40
- HI16-32-010-105
- HI16-32-010-15
- HI16-32-025-105
- HI16-53-050-50
- HI16-20-033-25
- HI16-32-010-60
- HI16-53-050-30
- HI16-20-033-30
- HI16-53-100-15
- HI16-53-010-30
- HI16-53-025-15
- HI16-25-050-105
- HI16-53-010-10
- HI16-53-010-15
- HI16-53-025-60
- HI16-18-040-20
- HI16-53-050-10
- HI16-15-015-5
- HI16-25-050-15
- HI16-15-015-30
- HI16-25-050-30
- HI16-18-040-60
- HI16-25-100-30
- HI16-53-050-15
- HI16-25-100-105
- HI16-25-100-50
- HI16-18-036-60
- HI16-32-150-60
- HI16-25-025-30G5
- HI16-25-025-60G5
- HI16-32-100-105
- HI16-25-025-15G5
- HI16-32-150-105
- HI16-25-010-60
- HI16-25-025-10
- HI16-53-100-30G5
- HI16-25-010-50
- HI16-32-150-10
- HI16-18-020-40
- HI16-25-025-50
- HI16-10-040-10
- HI16-25-025-105
- HI16-18-020-10
- HI16-15-015-20
- HI16-15-015-15
- HI16-18-020-50
- HI16-15-015-10
- HI16-25-025-60
- HI16-10-040-5
- HI16-25-050-10
- Phase name:
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- HI-1 MS
- Phase composition:
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- 100% Methyl Polysiloxane (100% Dimethylpolysiloxane) - low bleeding
- Polarity:
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Apolar stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- USP code:
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38
- G1, G2, G9, G38