Order Entry
Czech Republic
ContactUsLinkComponent
Thermal transfer ribbon for printer, R7942 series
Thermal transfer ribbon for printer, R7942 series
  BRDY237556
 :  Brady
undefined
Thermal transfer ribbon for printer, R7942 series
  BRDY237556
 :  Brady

 

  • Colour:
    Black
  • Description:
    Thermal transfer printer ribbon, R7942 series, ink on the side
  • Length:
    100 m
  • Environmentally Preferable:
  • Pk:
    1
  • For:
    BP1244, BP1344

 

 

Waxed ribbon for general use on paper and for temporary identification.

  • Width: 110 mm
 : One print ribbon/box.