Labels for BBP33 printers, type B-7642, PermaSleeve® HX
Supplier: Brady
Certificates
About this item
PermaSleeve® HX Polyolefin wire marker sleeves are military/industry grade.
- Flame-retardant
- Temperature resistance from –40 to 120 °C
Certifications: Meets SAE-AMS-DTL-23053/5 (class 1 and 3), SAE-AS-81531- MIL-STD-202 and tested ASTM E162, ASTM E662.
Frequently Bought Together