2-Acetyl-3-ethylpyrazine, Sigma-Aldrich®
Catalog # SIALW325007-100G-K
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:150.18 g/mol
:M-Clarity™ quality level = MQ300
EINECS:251-316-2
CAS Number:32974-92-8
Formula:C₈H₁₀N₂O
MDL Number:MFCD00038028