

1,3-Propanesultone, semi-solid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1120-71-4
:
Formula:C₃H₆O₃S
MDL Number:MFCD00005355
:122.14 g/mol
Boiling Pt:298 °C (1013 hPa)
Melting Pt:30…33 °C (lit.)
Density:1.392 g/ml
Storage Temperature:Refrigerator
EINECS:214-317-9
UN:2811
ADR:6.1,III
:GHS08
:GHS07
Frequently Bought Together