Tetrahydro-4H-pyran-4-one, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:29943-42-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00006581
:100.12 g/mol
Boiling Pt:166 °C (1013 hPa)
Density:1.08 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:249-967-2
Frequently Bought Together