2,6-Dimethyl-4-pyrone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1004-36-0
:
Formula:C₇H₈O₂
MDL Number:MFCD00006579
Boiling Pt:248 °C (1013 hPa)
Melting Pt:130…131 °C
Density:1.17 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
:124.14 g/mol
EINECS:213-719-1
Frequently Bought Together