Order Entry
Czech Republic
ContactUsLinkComponent
Ruthenium(III) acetylacetonate for synthesis, Sigma-Aldrich®
Ruthenium(III) acetylacetonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ruthenium(III) acetylacetonate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    230 - 235 °C
  • Molar Mass:
    398.39 g/mol
  • Synonyms:
    Tris(acetylacetonato)ruthenium(III), Tris(pentane-2,4-dionato)ruthenium(III)
  • Hill Formula:
    C15H21O6Ru
  • Chemical Formula:
    (CH3C(O)=CHCOCH3)3Ru

Specification Test Results

Related Information

Frequently Bought Together