Order Entry
Czech Republic
ContactUsLinkComponent
 
Oxidizing
Palladium(II) oxide hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Catalog # 8.07107.0001
Supplier:  Merck
CAS Number:  
Oxidizing
Palladium(II) oxide hydrogenation catalyst, for synthesis, Sigma-Aldrich®
Catalog # 8.07107.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    122.42 g/mol
  • Hill Formula:
    OPd
  • Chemical Formula:
    PdO

Specification Test Results

About this item

 :
 :122.42 g/mol
Merck Index:12,07125
Melting Pt:870 °C
Density:8.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-218-3
UN:1479
ADR:5.1,II
 :M-Clarity™ quality level = MQ200
 :GHS03
CAS Number:1314-08-5
Formula:OPd
MDL Number:MFCD00011172