Palladium(II) oxide, anhydrous ≥99.9% (metals basis) Pd ≥ 85%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-08-5
:
Formula:OPd
MDL Number:MFCD00011172
:122.42 g/mol
Merck Index:14,6993
Melting Pt:870 °C
Density:8.70
Storage Temperature:Ambient
EINECS:215-218-3
Related Information
Frequently Bought Together