
Silver(I) hexafluorophosphate 98% -6 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:26042-63-7
:
Formula:AgF₆P
MDL Number:MFCD00003415
:252.83
Storage Temperature:Ambient
EINECS:247-428-6
UN:1759
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to grey-beige Powder
- Silver (Ag):≥41.6 %
Specification Test Results
Frequently Bought Together