
2-Acetylnaphthalene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:93-08-3
:
Formula:C₁₂H₁₀O
MDL Number:MFCD00004108
:170.21 g/mol
Boiling Pt:300 °C (1013 hPa)
Melting Pt:52…54 °C
Density:1.1171 g/cm³ (20 °C)
EINECS:202-216-2
ADR:9,III
:M-Clarity™ quality level = MQ200
UN:3077
:GHS09
Storage Temperature:Ambient
Frequently Bought Together