
1-Nitroso-2-naphthol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:131-91-9
:
Formula:C₁₀H₇NO₂
MDL Number:MFCD00003884
:173.17 g/mol
Merck Index:14,6641
Boiling Pt:304 °C (1013 hPa)
Melting Pt:104…108 °C (dec.)
Storage Temperature:Ambient
EINECS:205-043-0
ADR:9,III
:GHS07
Related Information
Frequently Bought Together