

1-Nitroso-2-naphthol 98%
:131-91-9
:
:C₁₀H₇NO₂
:MFCD00003884
:173.17 g/mol
:12,06739
:304 °C (1013 hPa)
:104…108 °C
:Ambient
:205-043-0
:3077
:GHS07
:GHS09
- Appearance:Brown Powder
- Infrared spectrum:Conforms
- Melting point:104 to 108 °C
- Water:≤0.5 %
- Titration with NaOH:97.5 to 102.5 % (On dry substance)
Frequently Bought Together