

N,N-Dimethyl-1-naphthylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-56-6
:
Formula:C₁₂H₁₃N
MDL Number:MFCD00003919
:171.24 g/mol
Merck Index:12,03301
Boiling Pt:139…140 °C (13 mmHg)
Density:1.04 g/cm³
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:201-682-4
UN:3082
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear light yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6210 to 1.6230 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together