4-Chloro-1-naphthol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:604-44-4
:
Formula:ClC₁₀H₆OH
MDL Number:MFCD00003974
:178.62 g/mol
Boiling Pt:307 °C (1013 hPa)
Melting Pt:123 °C
Density:1.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-068-5
Frequently Bought Together