
4-Chloro-1-naphthol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:604-44-4
:
Formula:ClC₁₀H₆OH
MDL Number:MFCD00003974
:178.62 g/mol
Boiling Pt:307 °C (1013 hPa)
Melting Pt:117…120 °C
Density:1,2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-068-5
:GHS07
Specification Test Results
- Appearance:Off-white to grey to light brown Powder or needles
- Infrared spectrum:Conforms
- Melting point:117 to 122 °C
- GC:≥96.0 %
- UV:at 280 nm A: ≤0.1
- Titration after Ion exchange:≥99.0 %
Specification Test Results
Frequently Bought Together