Order Entry
Czech Republic
ContactUsLinkComponent
Praseodymium trinitrate pentahydrate 99.9% (trace metals basis)
Oxidizing
Praseodymium trinitrate pentahydrate 99.9% (trace metals basis)
Catalog # ACRO194590500
CAS Number:  
undefined
Oxidizing
Praseodymium trinitrate pentahydrate 99.9% (trace metals basis)
Catalog # ACRO194590500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Appearance:
    Green Crystalline powder
  • Total conc. of imp.as REO:
    ≤0.1 %

Specification Test Results

About this item

 :
 :417
Melting Pt:69 °C
Density:2,233 g/cm³ (20 °C)
EINECS:233-796-5
UN:1477
ADR:5.1,II
 :GHS03
CAS Number:14483-17-1
Formula:N₃O₉Pr·5H₂O