Di-tert-butyl disulphide for synthesis, Sigma-Aldrich®
Catalog # 8.20243.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-5 °C
- Molar Mass:178.35 g/mol
- Boiling Point:198.2 °C
- Vapor Pressure:2 hPa (38 °C)
- Flash Point:67 °C
- Explosion Limit:0.8 %(V)
- Density:0.93 g/cm3 (20 °C)
Specification Test Results
About this item
:
:178.36 g/mol
Boiling Pt:198 °C (1013 hPa)
Melting Pt:–5 °C
Density:0.9229 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-734-1
CAS Number:110-06-5
Formula:C₈H₁₈S₂
MDL Number:MFCD00008838