

Di-tert-butyl disulphide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-06-5
:
Formula:C₈H₁₈S₂
MDL Number:MFCD00008838
:178.36 g/mol
Boiling Pt:198…204 °C (760 mmHg)
Melting Pt:–5 °C
Density:0.93 g/cm³
Flash Pt:67 °C
Storage Temperature:Ambient
EINECS:203-734-1
UN:3334
ADR:9,III
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4910 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together