
Ammonium metavanadate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7803-55-6
:MFCD00011453
Formula:NH₄VO₃
MDL Number:MFCD00011430
:116.98 g/mol
Merck Index:13,00572
Melting Pt:200 °C
Density:2.33 g/cm³ (20 °C)
EINECS:232-261-3
ADR:6.1,II
:M-Clarity™ quality level = MQ200
UN:2859
:GHS06
Frequently Bought Together