Zadání objednávky
Czech Republic
ContactUsLinkComponent
Metavanadičnan amonný Reag. Ph. Eur. analytical reagent, Supelco®
Toxic
Health
Environment
Metavanadičnan amonný Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates
Toxic
Health
Environment
Metavanadičnan amonný Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    5.1 g/l (20 °C)
  • Melting Point:
    200 °C (decomposition)
  • Molar Mass:
    116.97 g/mol
  • Bulk Density:
    1000 kg/m3
  • Density:
    2.3 g/cm3 (20 °C)
  • pH:
    7 (5.1 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together